Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172063.2 | NP_001165534.1 | 8 - 9 | 1600 | 91 | |
XM_006242294.2 | XP_006242356.1 | 10 - 11 | 2686 | 91 | ||
XM_006242295.2 | XP_006242357.1 | 10 - 11 | 2009 | 91 | ||
XM_006242296.2 | XP_006242358.1 | 10 - 11 | 1953 | 91 | ||
XM_008765651.1 | XP_008763873.1 | 11 - 12 | 2188 | 91 | ||
XM_008765652.1 | XP_008763874.1 | 10 - 11 | 1945 | 91 | ||
XM_008765653.1 | XP_008763875.1 | 10 - 11 | 2225 | 91 | ||
GenBank mRNA | FQ211776.1 | - | 10 - 11 | 1977 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |