Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E2f4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271345.1 | NP_001258274.1 | 8 - 9 | 1164 | 91 | |
XM_006255433.1 | XP_006255495.1 | 8 - 9 | 1090 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |