Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces2h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044258.2 | NP_001037723.1 | 2 - 3 | 316 | 103 | |
NM_001190346.1 | NP_001177275.1 | 2 - 3 | 316 | 103 | ||
XM_008772523.1 | XP_008770745.1 | 2 - 3 | 335 | 103 | ||
XM_008772524.1 | XP_008770746.1 | 3 - 4 | 458 | 103 | ||
GenBank mRNA | BC107806.1 | - | 2 - 3 | 286 | 7368719 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |