Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195558.1 | NP_001182487.1 | 3 - 4 | 945 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Rattus norvegicus Probe spans exons |