Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bace2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002802.1 | NP_001002802.1 | 4 - 5 | 735 | 150 | |
XM_006248153.2 | XP_006248215.1 | 4 - 5 | 789 | 150 | ||
GenBank mRNA | BC166597.1 | - | 4 - 5 | 749 | 7936295 | 150 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Rattus norvegicus Probe spans exons |