Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hlcs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006248111.2 | XP_006248173.1 | 4 - 5 | 2286 | 111 | |
XM_006248112.2 | XP_006248174.1 | 4 - 5 | 2353 | 111 | ||
XM_006248114.2 | XP_006248176.1 | 3 - 4 | 2147 | 111 | ||
XM_008768611.1 | XP_008766833.1 | 4 - 5 | 1485 | 111 | ||
XM_008768612.1 | XP_008766834.1 | 3 - 4 | 1350 | 111 | ||
XM_221630.8 | XP_221630.6 | 3 - 4 | 1466 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |