Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Siglec10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106250.1 | NP_001099720.1 | 6 - 7 | 1628 | 84 | |
XM_006228948.2 | XP_006229010.1 | 9 - 10 | 1787 | 84 | ||
GenBank mRNA | BC168854.1 | - | 4 - 5 | 861 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |