Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100831.1 | NP_001094301.1 | 2 - 3 | 472 | 86 | |
GenBank mRNA | BC105894.1 | - | 2 - 3 | 237 | 86 | |
CD371338.1 | - | 2 - 3 | 482 | 86 | ||
FQ213223.1 | - | 2 - 3 | 272 | 86 | ||
FQ214314.1 | - | 2 - 3 | 269 | 86 | ||
FQ223219.1 | - | 2 - 3 | 315 | 86 | ||
FQ233693.1 | - | 2 - 3 | 303 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |