Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ceacam20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170324.1 | NP_001163795.1 | 4 - 5 | 730 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus Probe spans exons |