Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001076463.5 | XP_001076463.2 | 1 - 2 | 146 | 60 | |
XM_008759458.1 | XP_008757680.1 | 1 - 2 | 82 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |