Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfkbid | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003748825.3 | XP_003748873.1 | 8 - 9 | 1299 | 79 | |
XM_006228804.2 | XP_006228866.1 | 7 - 8 | 1785 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |