Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfkbid | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003748825.3 | XP_003748873.1 | 10 - 11 | 1570 | 53 | |
XM_006228804.2 | XP_006228866.1 | 9 - 10 | 2056 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |