Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025678.1 | NP_001020849.1 | 1 - 2 | 50 | 135 | |
XM_006228414.2 | XP_006228476.1 | 2 - 3 | 2132 | 135 | ||
XM_006228415.2 | XP_006228477.1 | 2 - 3 | 2135 | 135 | ||
GenBank mRNA | BC099170.1 | - | 1 - 2 | 50 | 7382397 | 135 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |