Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stoml2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031646.1 | NP_001026816.1 | 8 - 9 | 806 | 81 | |
GenBank mRNA | BC099164.1 | - | 8 - 9 | 806 | 7382023 | 81 |
FQ213146.1 | - | 8 - 9 | 837 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |