Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aasdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006250858.2 | XP_006250920.1 | 10 - 11 | 2102 | 102 | |
XM_008770109.1 | XP_008768331.1 | 7 - 8 | 1361 | 102 | ||
XM_008770110.1 | XP_008768332.1 | 7 - 8 | 1361 | 102 | ||
XM_008770111.1 | XP_008768333.1 | 7 - 8 | 1361 | 102 | ||
XM_008770112.1 | XP_008768334.1 | 2 - 3 | 1492 | 102 | ||
XM_008770113.1 | XP_008768335.1 | 3 - 4 | 451 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |