Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brdt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012031.2 | NP_001012031.1 | 5 - 6 | 821 | 77 | |
GenBank mRNA | BC078999.1 | - | 5 - 6 | 799 | 77 | |
BC158630.1 | - | 5 - 6 | 796 | 7456383 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |