Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106815.1 | NP_001100285.1 | 7 - 8 | 1740 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |