Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubash3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191792.1 | NP_001178721.1 | 2 - 3 | 221 | 103 | |
XM_006242848.2 | XP_006242910.1 | 2 - 3 | 546 | 103 | ||
XM_006242850.2 | XP_006242912.1 | 2 - 3 | 795 | 103 | ||
GenBank mRNA | FQ223998.1 | - | 2 - 3 | 472 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |