Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pus7l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002729847.4 | XP_002729893.2 | 4 - 5 | 1366 | 72 | |
XM_006242271.2 | XP_006242333.1 | 4 - 5 | 1673 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |