Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdelr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127546.1 | NP_001121018.1 | 4 - 5 | 671 | 98 | |
XM_008765714.1 | XP_008763936.1 | 4 - 5 | 1389 | 98 | ||
GenBank mRNA | BC162033.1 | - | 4 - 5 | 671 | 9103157 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |