Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rangap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244855.1 | NP_001231784.1 | 11 - 12 | 1470 | 92 | |
XM_006242100.2 | XP_006242162.1 | 11 - 12 | 2910 | 92 | ||
XM_006242101.2 | XP_006242163.1 | 11 - 12 | 1759 | 92 | ||
GenBank mRNA | FQ217036.1 | - | 11 - 12 | 1454 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |