Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rangap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244855.1 | NP_001231784.1 | 12 - 13 | 1591 | 86 | |
XM_006242100.2 | XP_006242162.1 | 12 - 13 | 3031 | 86 | ||
XM_006242101.2 | XP_006242163.1 | 12 - 13 | 1880 | 86 | ||
GenBank mRNA | BC082056.1 | - | 12 - 13 | 1499 | 7132303 | 86 |
BP492086.1 | - | 2 - 3 | 142 | 86 | ||
FQ217036.1 | - | 12 - 13 | 1575 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |