Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpnpep3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130582.2 | NP_001124054.2 | 8 - 9 | 1289 | 66 | |
XM_006242121.2 | XP_006242183.1 | 7 - 8 | 1106 | 66 | ||
GenBank mRNA | BC168759.1 | - | 8 - 9 | 1289 | 6888763 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Rattus norvegicus Probe spans exons |