Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271215.1 | NP_001258144.1 | 3 | 300 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Amplicon spans exons and probe does not span exons |