Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pknox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013074.1 | NP_001013092.1 | 1 - 2 | 126 | 83 | |
XM_006256187.2 | XP_006256249.1 | 2 - 3 | 1161 | 83 | ||
XM_006256188.2 | XP_006256250.1 | 3 - 4 | 1223 | 83 | ||
XM_006256189.2 | XP_006256251.1 | 3 - 4 | 1215 | 83 | ||
XM_006256190.2 | XP_006256252.1 | 2 - 3 | 1162 | 83 | ||
XM_008772808.1 | XP_008771030.1 | 3 - 4 | 1038 | 83 | ||
XM_008772809.1 | XP_008771031.1 | 2 - 3 | 1153 | 83 | ||
XM_008772810.1 | XP_008771032.1 | 2 - 3 | 1041 | 83 | ||
GenBank mRNA | BC091397.1 | - | 1 - 2 | 126 | 7323315 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |