Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srp54a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053871.1 | NP_446323.1 | 14 - 15 | 1565 | 99 | |
GenBank mRNA | BC079117.1 | - | 14 - 15 | 1565 | 7126297 | 99 |
FQ211580.1 | - | 14 - 15 | 1576 | 99 | ||
FQ225417.1 | - | 14 - 15 | 1580 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |