Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc39a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750698.3 | XP_003750746.1 | 6 - 7 | 4979 | 59 | |
XM_006244936.2 | XP_006244998.1 | 6 - 7 | 1915 | 59 | ||
XM_006244937.2 | XP_006244999.1 | 7 - 8 | 1903 | 59 | ||
XM_006244938.2 | XP_006245000.1 | 6 - 7 | 2933 | 59 | ||
XM_008767152.1 | XP_008765374.1 | 6 - 7 | 2491 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter Recommended Assay Rattus norvegicus Inventoried Probe spans exons |