Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ms4a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749066.3 | XP_003749114.1 | 2 - 3 | 414 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |