Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012089.1 | NP_001012089.1 | 2 - 3 | 583 | 85 | |
GenBank mRNA | BC088205.1 | - | 2 - 3 | 583 | 7380627 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |