Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oog1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006239344.2 | XP_006239406.1 | 3 - 4 | 988 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |