Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspyl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191618.2 | NP_001178547.1 | 2 - 3 | 994 | 78 | |
XM_006257038.2 | XP_006257100.1 | 2 - 3 | 1145 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |