Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nutm2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008771552.1 | XP_008769774.1 | 4 - 5 | 3085 | 123 | |
XM_225188.7 | XP_225188.5 | 5 - 6 | 1438 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |