Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec2d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085402.1 | NP_001078871.1 | 1 - 2 | 165 | 104 | |
GenBank mRNA | EF100686.1 | - | 1 - 2 | 165 | 104 | |
EU128746.1 | - | 1 - 2 | 171 | 104 | ||
FQ221935.1 | - | 256 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus Probe spans exons |