Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgals5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012976.2 | NP_037108.1 | 3 - 4 | 324 | 149 | |
GenBank mRNA | FQ225275.1 | - | 3 - 4 | 318 | 149 | |
FQ225984.1 | - | 3 - 4 | 340 | 149 | ||
FQ227276.1 | - | 3 - 4 | 339 | 149 | ||
FQ228081.1 | - | 3 - 4 | 344 | 149 | ||
FQ230182.1 | - | 3 - 4 | 344 | 149 | ||
FQ231538.1 | - | 3 - 4 | 343 | 149 | ||
FQ231564.1 | - | 3 - 4 | 354 | 149 | ||
FQ232247.1 | - | 3 - 4 | 365 | 149 | ||
FQ232513.1 | - | 3 - 4 | 347 | 149 | ||
FQ233614.1 | - | 3 - 4 | 365 | 149 | ||
FQ234601.1 | - | 3 - 4 | 347 | 149 | ||
FQ234845.1 | - | 3 - 4 | 367 | 149 | ||
L36862.1 | - | 3 - 4 | 324 | 149 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |