Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk1c10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135173.1 | NP_001128645.1 | 191 | 136 | ||
GenBank mRNA | S48142.1 | - | 1 - 2 | 148 | 136 |
Gene Symbol: LOC100911689 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003748848.3 | XP_003748896.1 | 211 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Protease Rattus norvegicus Probe spans exons |