Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hbe2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024805.1 | NP_001019976.1 | 2 - 3 | 311 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |