Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191902.1 | NP_001178831.1 | 2 | 443 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon |