Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acot2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138907.2 | NP_620262.2 | 3 | 2139 | 130 | |
GenBank mRNA | BC072540.1 | - | 3 | 2139 | 7104319 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon |