Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009175.1 | NP_001009175.1 | 3 - 4 | 658 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |