Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ebag9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009665.1 | NP_001009665.1 | 4 - 5 | 532 | 136 | |
XM_006241613.2 | XP_006241675.1 | 4 - 5 | 907 | 136 | ||
XM_008765451.1 | XP_008763673.1 | 3 - 4 | 620 | 136 | ||
XM_008765452.1 | XP_008763674.1 | 4 - 5 | 3570 | 136 | ||
GenBank mRNA | BC087093.1 | - | 4 - 5 | 532 | 7191226 | 136 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |