Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom1r49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008920.1 | NP_001008920.1 | 852 | 129 | ||
GenBank mRNA | AY510303.1 | - | 852 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Recommended Assay Rattus norvegicus Inventoried |