Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom1r56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008929.1 | NP_001008929.1 | 1 | 672 | 177 | |
GenBank mRNA | AY510280.1 | - | 1 | 672 | 177 |
Gene Symbol: LOC100909792 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749304.2 | XP_003749352.1 | 1 | 725 | 177 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Rattus norvegicus Inventoried Both primers and probe map within a single exon |