Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tGap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007635.2 | NP_001007636.2 | 20 - 21 | 2877 | 94 | |
XM_006232134.2 | XP_006232196.1 | 22 - 23 | 3219 | 94 | ||
GenBank mRNA | AY631396.1 | - | 20 - 21 | 2877 | 94 | |
AY631398.1 | - | 13 - 14 | 2021 | 94 | ||
AY631399.1 | - | 20 - 21 | 2642 | 94 | ||
AY631400.1 | - | 16 - 17 | 2016 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |