Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom2r56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099484.1 | NP_001092954.1 | 1548 | 127 |
Gene Symbol: LOC103690080 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008765207.1 | XP_008763429.1 | 4 - 5 | 710 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |