Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsbp1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136183.1 | NP_001129655.1 | 3 - 4 | 305 | 77 | |
GenBank mRNA | CR474979.1 | - | 1 - 2 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Rattus norvegicus 1700 Probe spans exons |