Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spred3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173344.1 | NP_001166815.1 | 2 - 3 | 781 | 96 | |
XM_008759144.1 | XP_008757366.1 | 3 - 4 | 419 | 96 | ||
XM_008759145.1 | XP_008757367.1 | 3 - 4 | 366 | 96 | ||
XM_008759146.1 | XP_008757368.1 | 3 - 4 | 488 | 96 | ||
GenBank mRNA | BC166515.1 | - | 2 - 3 | 781 | 7131550 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |