Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oxct2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012221.1 | NP_001012221.1 | 1 | 652 | 67 | |
GenBank mRNA | BC083681.1 | - | 1 | 652 | 7135892 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Both primers and probe map within a single exon |