Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom2r52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099503.1 | NP_001092973.1 | 2 - 3 | 526 | 76 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Recommended Assay Rattus norvegicus Probe spans exons |