Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkx2-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127653.1 | NP_001121125.1 | 1 - 2 | 67 | 92 | |
GenBank mRNA | BC160896.1 | - | 1 - 2 | 67 | 7453070 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons |