Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022511.2 | NP_071956.2 | 2 - 3 | 444 | 82 | |
GenBank mRNA | BC062405.1 | - | 2 - 3 | 444 | 5623232 | 82 |
FQ217736.1 | - | 2 - 3 | 478 | 82 | ||
FQ221987.1 | - | 2 - 3 | 478 | 82 | ||
FQ227505.1 | - | 2 - 3 | 479 | 82 | ||
FQ228171.1 | - | 2 - 3 | 478 | 82 | ||
FQ232047.1 | - | 2 - 3 | 477 | 82 | ||
FQ232161.1 | - | 2 - 3 | 434 | 82 | ||
FQ232713.1 | - | 2 - 3 | 448 | 82 | ||
FQ235292.1 | - | 2 - 3 | 479 | 82 | ||
X96967.1 | - | 2 - 3 | 327 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |